Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terephthalic-d4 acid, for NMR, 98% atom D, Acros Organics

Click to view available options
Quantity:
1g
60088-54-2
C8H6O4
170.156
MFCD00002559
KKEYFWRCBNTPAC-RHQRLBAQSA-N
terephthalic-d4 acid,2,3,5,6-2h4 terephthalic acid,terephthalic-2,3,5,6-d4 acid,terephthalic-d4,terephthalic-d4-acid,1,4-benzene-d4-dicarboxylic acid,2 h? benzene-1,4-dicarboxylic acid,terephthalic-2,3,5,6-d4 acid 98 atom % d cp,terephthalic-2,3,5,6-d4 acid, 98 atom % d cp
108885
2,3,5,6-tetradeuterioterephthalic acid
C1=CC(=CC=C1C(=O)O)C(=O)O
60088-54-2
C8H6O4
170.156
MFCD00002559
KKEYFWRCBNTPAC-RHQRLBAQSA-N
terephthalic-d4 acid,2,3,5,6-2h4 terephthalic acid,terephthalic-2,3,5,6-d4 acid,terephthalic-d4,terephthalic-d4-acid,1,4-benzene-d4-dicarboxylic acid,2 h? benzene-1,4-dicarboxylic acid,terephthalic-2,3,5,6-d4 acid 98 atom % d cp,terephthalic-2,3,5,6-d4 acid, 98 atom % d cp
108885
2,3,5,6-tetradeuterioterephthalic acid
C1=CC(=CC=C1C(=O)O)C(=O)O
Color White
Deuterium (D) 97.5% min.
Quantity 1g
Linear Formula C6D4(CO2H)2
Formula Weight 170.17
Physical Form Powder or Needles
Grade NMR
Chemical Name or Material Terephthalic-d4 acid, 98 atom % D