Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERF1 Mouse anti-Human, Clone: BED5, Abnova™

Mouse monoclonal antibody raised against human TERF1.

Manufacturer:  Abnova Corporation MAB12646

Catalog No. 89-934-990


Add to cart

Description

Description

This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products. [provided by RefSeq]

Specifications

Specifications

TERF1
BED5
Mouse monoclonal antibody raised against human TERF1.
In PBS (0.09% sodium azide).
FLJ41416/PIN2/TRBF1/TRF/TRF1/hTRF1-AS/t-TRF1
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed)
Unconjugated
ELISA Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TERF1
Recombinant protein corresponding to human TERF1.
Affinity purification
RUO
Primary
7013
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit