Learn More
TERF2IP Antibody (78B356.1), Novus Biologicals™
SDP
Mouse Monoclonal Antibody has been used in 2 publications
Supplier:  Novus Biologicals NB10056526
 View more versions of this product
Catalog No. NB10056526
Only null left
Add to Cart
Description
Description
TERF2IP Monoclonal specifically detects TERF2IP in Human samples. It is validated for Western Blot.
Specifications
Specifications
TERF2IP
Monoclonal
0.5 mg/mL
Western Blot 0.1-1 ug/ml
dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein
Mouse
Protein G purified
RUO
Primary
Human
Purified
Western Blot
78B356.1
Unconjugated
Q9NYB0
TERF2IP
A synthetic peptide corresponding amino acids 212-235 of human Rap1 was used to immunize mice.
0.1 mg
Chromatin Research, DNA Repair, Endosome Markers
54386
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only