Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TERF2IP Antibody (78B356.1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 2 publications

$206.50 - $499.50

Specifications

Antigen TERF2IP
Clone 78B356.1
Concentration 0.5 mg/mL
Dilution Western Blot 0.1-1 ug/ml
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056526T
SDP
View Documents
Novus Biologicals
NB10056526SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056526
SDP
View Documents
Novus Biologicals
NB10056526
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

TERF2IP Monoclonal specifically detects TERF2IP in Human samples. It is validated for Western Blot.
Specifications

Specifications

TERF2IP
0.5 mg/mL
Western Blot
Unconjugated
Mouse
Chromatin Research, DNA Repair, Endosome Markers
Q9NYB0
54386
A synthetic peptide corresponding amino acids 212-235 of human Rap1 was used to immunize mice.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
78B356.1
Western Blot 0.1-1 ug/ml
Monoclonal
Purified
RUO
Human
dopamine receptor interacting protein 5, Dopamine receptor-interacting protein 5, DRIP5, hRap1, RAP1 homolog, RAP1TERF2-interacting telomeric protein 1, Repressor/activator protein 1 homolog, telomeric repeat binding factor 2, interacting protein, telomeric repeat-binding factor 2-interacting protein 1, TRF2-interacting telomeric protein 1, TRF2-interacting telomeric RAP1 protein
TERF2IP
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.