Learn More
Tergitol(R) 15-S-5, Surfactant, Spectrum™
T1255, 84133-50-6, C12-14H25-29O(CH2CH2O)xH
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner T1255100ML
Catalog No. 18-607-249
Add to cart
Description
Description
Spectrum™ Tergitol(R) 15-S-5, Surfactant is a nonionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.
Specifications
Specifications
100 mL