Learn More
Tergitol(R) 15-S-7, Surfactant, Spectrum™
T1258, 84133-50-6, C12-14H25-29O[CH2CH2O]xH
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner T125820LT
 View more versions of this product
Catalog No. 18-607-257
Add to cart
Description
Description
Spectrum™ Tergitol(R) 15-S-7, Surfactant is a Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.
Specifications
Specifications
C12-14-H25-29[CheCheO]xH
5.5 - 7.5
20 L