Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tergitol(R) NP-10, Surfactant, Spectrum™

Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
127087-87-0
127087-87-0

T1273, 127087-87-0, C15H24O(C2H4O)x

Spectrum™ Tergitol(R) NP-10, Surfactant is a Nonylphenol Ethoxylate based surfactant. It is nonionic and provides excellent detergency, rinse-ability, and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and metalworking fluids.

CAS Max % 1
Packaging Steel Can
Grade Reagent
Water 0.003
pH 5.0 to 8.0
Quantity 20 L