Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tergitol(R) NP-4, Surfactant, Spectrum™

Click to view available options
Quantity:
100 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Pail
127087-87-0
127087-87-0

T1276, 127087-87-0, C15H24O(C2H4O)x

Spectrum™ Tergitol(R) NP-4, Surfactant is a Nonylphenol Ethoxylate surfactant. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals.

CAS Max % 1
Packaging Steel Pail
Grade Reagent
Water 0.005
pH 5.0 to 8.0
Quantity 20 L