Learn More
Tergitol(R) TMN-10, Surfactant, Spectrum™
T1288, 60828-78-6, C12H25O(CH2CH2O)nH
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner T12884LT
 View more versions of this product
Catalog No. 18-607-286
Add to cart
Description
Description
Spectrum™ Tergitol(R) TMN-10, Surfactant is a Nonylphenol Ethoxylate. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals.
Specifications
Specifications
C12H25O(CH2CH2O)nH
5-8
4 L