Learn More
Tergitol(R) TMN-6, Surfactant, Spectrum™
T1303, 60828-78-6, C12H25O(CH2CH2O)nH
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner T13035GLBL
 View more versions of this product
Catalog No. 18-607-291
Add to cart
Description
Description
Spectrum™ Tergitol(R) TMN-6, Surfactant is a is a water-soluble, nonionic surfactant that is a superior wetting agent for use in multiple applications including paper & textile processing and hard surface cleaners.
Specifications
Specifications
C12H26O(CH2CH2O)n (for Polyethylene glycol timethylnonyl ether)
106°C (223°F)
5 gal.