Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Terpinen-4-ol, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
562-74-3
C10H18O
154.25
MFCD00001562
WRYLYDPHFGVWKC-UHFFFAOYSA-N
terpinen-4-ol, 4-carvomenthenol, 4-terpineol, p-menth-1-en-4-ol, 1-terpinen-4-ol, terpinenol-4, terpene-4-ol, 1-menthene-4-ol, 1-p-menthen-4-ol, terpinene-4-ol
11230
CHEBI:78884
4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
CC1=CCC(CC1)(C(C)C)O
562-74-3
C10H18O
154.25
MFCD00001562
WRYLYDPHFGVWKC-UHFFFAOYSA-N
terpinen-4-ol, 4-carvomenthenol, 4-terpineol, p-menth-1-en-4-ol, 1-terpinen-4-ol, terpinenol-4, terpene-4-ol, 1-menthene-4-ol, 1-p-menthen-4-ol, terpinene-4-ol
11230
CHEBI:78884
4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
CC1=CCC(CC1)(C(C)C)O
Specific Gravity 0.929
Refractive Index 1.4760 to 1.4820
Flash Point 82°C
Quantity 100g
Specific Rotation -1° to +1° (20°C, 589nm) (neat)
Packaging Glass bottle
Infrared Spectrum Authentic
Density 0.9290g/mL
Boiling Point 212.0°C
Color Colorless to Yellow
Assay Percent Range 96% min. (GC)
Beilstein 06, 55
Merck Index 10, 8996
Solubility Information Solubility in water: very slightly soluble. Other solubilities: soluble in alcohol
Formula Weight 154.25
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Terpinen-4-ol, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-235-5

RTECSNumber : OT0175110

TSCA : TSCA