Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Terpineol (Laboratory), Fisher Chemical

Catalog Number T311
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
98-55-5
C10H18O
154.253
MFCD00001557
WUOACPNHFRMFPN-UHFFFAOYSA-N
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
17100
CHEBI:22469
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
CC1=CCC(CC1)C(C)(C)O
98-55-5
C10H18O
154.253
MFCD00001557
WUOACPNHFRMFPN-UHFFFAOYSA-N
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
17100
CHEBI:22469
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
CC1=CCC(CC1)C(C)(C)O

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY
Alpha Vector Terpineol
Melting Point 31°C
Density 0.937g/cm³
Boiling Point 214°C
Flash Point 90°C
Packaging Glass Bottle
Linear Formula CH3C:CHCH2CH[C(CH3)2OH]CH2CH2
Quantity 1 L
Vapor Pressure 0.05mmHg at 25°C
Formula Weight 154.25
Grade Laboratory
Physical Form Low Melting Solid, Solid
Chemical Name or Material α-Terpineol
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible material. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:2
Flammability:2
Instability:0
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit