Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Amylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
Glass bottle
594-39-8
C5H13N
87.166
MFCD00008056
GELMWIVBBPAMIO-UHFFFAOYSA-N
tert-amylamine,tert-pentylamine,2-butanamine, 2-methyl,t-amylamine,1,1-dimethylpropylamine,2-methyl-2-butylamine,2-amino-2-methylbutane,2-methyl-butan-2-amine,1,1-dimethyl-1-propylamine,t-pentylamine
68986
CHEBI:84236
2-methylbutan-2-amine
CCC(C)(C)N
594-39-8
C5H13N
87.166
MFCD00008056
GELMWIVBBPAMIO-UHFFFAOYSA-N
tert-amylamine,tert-pentylamine,2-butanamine, 2-methyl,t-amylamine,1,1-dimethylpropylamine,2-methyl-2-butylamine,2-amino-2-methylbutane,2-methyl-butan-2-amine,1,1-dimethyl-1-propylamine,t-pentylamine
68986
CHEBI:84236
2-methylbutan-2-amine
CCC(C)(C)N
Specific Gravity 0.746
Density 0.7460g/mL
Boiling Point 77.0°C
Refractive Index 1.3986 to 1.4006
Infrared Spectrum Authentic
Quantity 100mL
Flash Point −1°C
Packaging Glass bottle
Color Brown to Yellow
Assay Percent Range 98%
Linear Formula C2H5C(CH3)2NH2
Formula Weight 87.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material tert-Amylamine, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger