Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butanol, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Plastic bottle
75-65-0
C4H10O
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O
75-65-0
C4H10O
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol,tert-butyl alcohol,2-methyl-2-propanol,t-butanol,t-butyl hydroxide,1,1-dimethylethanol,trimethylcarbinol,trimethyl methanol,2-propanol, 2-methyl,t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O
Assay Percent Range 99.5%
Linear Formula (CH3)3COH
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone
Formula Weight 74.12
Percent Purity 99.5%
Grade Analysis
Density 0.7800g/mL
Packaging Plastic bottle
Refractive Index 1.3860 to 1.3880
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone
Specific Gravity 0.78
Flash Point 11°C
Viscosity 3.35 mPa.s (30°C)
Boiling Point 83.0°C
Melting Point 25.0°C to 25.5°C
Quantity 1L
Chemical Name or Material tert-Butanol
Show More Show Less