Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butoxybis(dimethylamino)methane, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5815-08-7
C9H22N2O
174.29
MFCD00042858
HXRAMSFGUAOAJR-UHFFFAOYSA-P
tert-butoxy bis dimethylamino methane,1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine,tert-butoxybis dimethylamino methane,bredereck's reagent,bredereck reagent,tert-butoxy dimethylamino methyl dimethylamine,unii-jej0u558f7,bis dimethylamino-t-butoxymethane,tert-butoxy-bis dimethylamino methane,bis dimethylamino tert-butoxy methane
79885
N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine
CC(C)(C)OC(N(C)C)N(C)C
5815-08-7
C9H22N2O
174.29
MFCD00042858
HXRAMSFGUAOAJR-UHFFFAOYSA-P
tert-butoxy bis dimethylamino methane,1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine,tert-butoxybis dimethylamino methane,bredereck's reagent,bredereck reagent,tert-butoxy dimethylamino methyl dimethylamine,unii-jej0u558f7,bis dimethylamino-t-butoxymethane,tert-butoxy-bis dimethylamino methane,bis dimethylamino tert-butoxy methane
79885
N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine
CC(C)(C)OC(N(C)C)N(C)C
Alkylation reagent
Refractive Index 1.4220 to 1.425
Boiling Point 50°C to 55°C (15.0mmHg)
Flash Point 41°C
Specific Gravity 0.844
Density 0.8440g/mL
Color Yellow
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Assay Percent Range 85+%
Linear Formula (CH3)3COCH[N(CH3)2]2
Merck Index 15, 1373
Solubility Information Solubility in water: decomposes
Formula Weight 174.29
Physical Form Liquid
Percent Purity 90% (sum of 3 constituents) varying proportionally: Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane
Chemical Name or Material tert-Butoxybis(dimethylamino)methane
Show More Show Less

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 227-383-9