Alfa Aesar™ tert-Butoxybis(dimethylamino)methane, tech. 90%

Manufacturer: Alfa Aesar™ H2786114

Quantity 25g

 View more versions of this product

Catalog No. AAH2786114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5815-08-7
    Molecular Formula C9H22N2O
    Linear Formula (CH3)3COCH[N(CH3)2]2
    Formula Weight 174.3
    MDL Number MFCD00042858
    UN Number UN1993
    Synonym Bredereck's reagent; 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine
    Chemical Name or Material tert-Butoxybis(dimethylamino)methane
    Grade Technical Grade

    Specifications

    Formula Weight 174.3
    Linear Formula (CH3)3COCH[N(CH3)2]2
    MDL Number MFCD00042858
    CAS 5815-08-7
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 50° to 55°C (15mmHg)
    Flash Point 41°C (106°F)
    Quantity 25g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Molecular Formula C9H22N2O
    Synonym Bredereck's reagent; 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material tert-Butoxybis(dimethylamino)methane
    Assay 90%
    Grade Technical Grade

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit