Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
35000-38-5
C24H25O2P
376.44
MFCD00075545
ZWZUFQPXYVYAFO-UHFFFAOYSA-N
tert-butyl 2-triphenylphosphoranylidene acetate,tert-butoxycarbonylmethylene triphenylphosphorane,tert-butyl triphenylphosphoranylidene acetate,acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester,t-butoxycarbonylmethylene triphenylphosphorane,tert-butyl 2-triphenyl-??-phosphanylidene acetate,triphenylthoranylidene acetic acid 1,1-dimethylethyl ester,pubchem14371,tert-butyl 2-triphenyl,tert-butyl 2-triphenylylidene acetate
256127
tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
35000-38-5
C24H25O2P
376.44
MFCD00075545
ZWZUFQPXYVYAFO-UHFFFAOYSA-N
tert-butyl 2-triphenylphosphoranylidene acetate,tert-butoxycarbonylmethylene triphenylphosphorane,tert-butyl triphenylphosphoranylidene acetate,acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester,t-butoxycarbonylmethylene triphenylphosphorane,tert-butyl 2-triphenyl-??-phosphanylidene acetate,triphenylthoranylidene acetic acid 1,1-dimethylethyl ester,pubchem14371,tert-butyl 2-triphenyl,tert-butyl 2-triphenylylidene acetate
256127
tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
For C-C Bond Formation (Olefination)
Packaging Glass bottle
Quantity 5g
Color White
Melting Point 146.0°C to 151.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (C6H5)3P=CHCO2C(CH3)3
Formula Weight 376.44
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material (tert-Butoxycarbonylmethylene)triphenylphosphorane
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : Refrigerator +4°C