Learn More
(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%, ACROS Organics™
Description
Specifications
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Melting Point | 146.0°C to 151.0°C |
Quantity | 25g |
Color | White |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Linear Formula | (C6H5)3P=CHCO2C(CH3)3 |
Formula Weight | 376.44 |
Show More |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Recommended Storage : Refrigerator +4°C