Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl 2,2,2-trichloroacetimidate, 95%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Description
Alkylation reagent
Specifications
Specific Gravity | 1.222 |
Boiling Point | 64.0°C to 66.0°C (15.0mmHg) |
Flash Point | 55°C |
Quantity | 5g |
Infrared Spectrum | Authentic |
Color | Colorless to Yellow |
Melting Point | 20.0°C |
Density | 1.2220g/mL |
Packaging | Glass bottle |
Refractive Index | 1.4550 to 1.4570 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning