Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2,2,2-trichloroacetimidate, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
98946-18-0
C6H10Cl3NO
218.502
MFCD00077410
CQXDYHPBXDZWBA-UHFFFAOYSA-N
tert-butyl 2,2,2-trichloroacetimidate,tert-butyl trichloroacetimidate,tert-butyl-2,2,2-trichloroacetimidate,tert-butyl 2,2,2-trichloroethanecarboximidate,2,2,2-trichloroacetimidic acid tert-butyl ester,t-butyl trichloroacetimidate,t-butyl 2,2,2-trichloroacetimidate,tert-butyl2,2,2-trichloroacetimidate,ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester,1-tert-butoxy-2,2,2-trichloroethanimine
2734700
tert-butyl 2,2,2-trichloroethanimidate
CC(C)(C)OC(=N)C(Cl)(Cl)Cl
98946-18-0
C6H10Cl3NO
218.502
MFCD00077410
CQXDYHPBXDZWBA-UHFFFAOYSA-N
tert-butyl 2,2,2-trichloroacetimidate,tert-butyl trichloroacetimidate,tert-butyl-2,2,2-trichloroacetimidate,tert-butyl 2,2,2-trichloroethanecarboximidate,2,2,2-trichloroacetimidic acid tert-butyl ester,t-butyl trichloroacetimidate,t-butyl 2,2,2-trichloroacetimidate,tert-butyl2,2,2-trichloroacetimidate,ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester,1-tert-butoxy-2,2,2-trichloroethanimine
2734700
tert-butyl 2,2,2-trichloroethanimidate
CC(C)(C)OC(=N)C(Cl)(Cl)Cl
Applications
tert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.

Solubility
Soluble in organic solvents, cyclo hexane.

Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.
Melting Point 20°C to 22°C
Quantity 1g
Boiling Point 64°C to 66°C (15mmHg)
Flash Point 55°C (131°F)
Assay Percent Range 0.97
UN Number UN1993
Beilstein 1770049
Formula Weight 218.51
Percent Purity 97%
Chemical Name or Material tert-Butyl 2,2,2-trichloroacetimidate

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

storageNote1 : Moisture Sensitive

Recommended Storage : 2° to 8°C