Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl 3-oxocyclobutanecarboxylate, 97%

Click to view available options
Quantity:
250mg
1g
5g
145549-76-4
C9H14O3
170.208
MFCD12827541
JINYZTGTQXDUQR-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutanecarboxylate,3-oxo-cyclobutanecarboxylic acid tert-butyl ester,cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester,tert-butyl 3-oxo-cyclobutanecarboxylate,1-boc-cyclobutan-3-one,t-butyl 3-oxocyclobutanecarboxylate,3-tert-butyloxycarbonylcyclobutanone,tert-butyl3-oxocyclobutanecarboxylate,tert-butyl 3-oxocyclobutane carboxylate,t-butyl-3-oxocyclobutanecarboxylate
10261520
tert-butyl 3-oxocyclobutane-1-carboxylate
CC(C)(C)OC(=O)C1CC(=O)C1
145549-76-4
C9H14O3
170.208
MFCD12827541
JINYZTGTQXDUQR-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutanecarboxylate,3-oxo-cyclobutanecarboxylic acid tert-butyl ester,cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester,tert-butyl 3-oxo-cyclobutanecarboxylate,1-boc-cyclobutan-3-one,t-butyl 3-oxocyclobutanecarboxylate,3-tert-butyloxycarbonylcyclobutanone,tert-butyl3-oxocyclobutanecarboxylate,tert-butyl 3-oxocyclobutane carboxylate,t-butyl-3-oxocyclobutanecarboxylate
10261520
tert-butyl 3-oxocyclobutane-1-carboxylate
CC(C)(C)OC(=O)C1CC(=O)C1
Quantity 5g
Formula Weight 170.2
Percent Purity 97%
Chemical Name or Material tert-Butyl 3-oxocyclobutanecarboxylate

TSCA : No

Recommended Storage : Ambient temperatures