Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (3-oxocyclobutyl)carbamate, 95%, Thermo Scientific Chemicals

Catalog Number p-4380062
Click to view available options
Quantity:
1 g
5 g
154748-49-9
C9H15NO3
185.22
FNHPTFKSPUTESA-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutylcarbamate, tert-butyl 3-oxocyclobutyl carbamate, tert-butyl n-3-oxocyclobutyl carbamate, 3-oxo-cyclobutyl-carbamic acid tert-butyl ester, 3-boc-amino cyclobutanone, 2-methyl-2-propanyl 3-oxocyclobutyl carbamate, 3-tert-butyloxycarbonylamino cyclobutan-1-one, 3-oxocyclobutylcarbamic acid, 1,1-dimethylethyl ester, carbamic acid, 3-oxocyclobutyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-oxocyclobutyl-, 1,1-dimethylethyl ester
23374584
tert-butyl N-(3-oxocyclobutyl)carbamate
CC(C)(C)OC(=O)NC1CC(=O)C1
154748-49-9
C9H15NO3
185.22
FNHPTFKSPUTESA-UHFFFAOYSA-N
tert-butyl 3-oxocyclobutylcarbamate, tert-butyl 3-oxocyclobutyl carbamate, tert-butyl n-3-oxocyclobutyl carbamate, 3-oxo-cyclobutyl-carbamic acid tert-butyl ester, 3-boc-amino cyclobutanone, 2-methyl-2-propanyl 3-oxocyclobutyl carbamate, 3-tert-butyloxycarbonylamino cyclobutan-1-one, 3-oxocyclobutylcarbamic acid, 1,1-dimethylethyl ester, carbamic acid, 3-oxocyclobutyl-, 1,1-dimethylethyl ester, carbamic acid, n-3-oxocyclobutyl-, 1,1-dimethylethyl ester
23374584
tert-butyl N-(3-oxocyclobutyl)carbamate
CC(C)(C)OC(=O)NC1CC(=O)C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 122°C to 124°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Formula Weight 185.22
Percent Purity 95%
Quantity 1 g
Chemical Name or Material tert-Butyl (3-oxocyclobutyl)carbamate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit