Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl (3S)-3-amino-4-hexenoate, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass Bottle
204587-90-6
C10H19NO2
185.267
MFCD00798296
LHUYXHJTMVKEOM-MRVPVSSYSA-N
tert-butyl 3s-3-amino-4-hexenoate,tert-butyl 3s-3-aminohex-4-enoate
57375595
tert-butyl (3S)-3-aminohex-4-enoate
CC=CC(CC(=O)OC(C)(C)C)N
204587-90-6
C10H19NO2
185.267
MFCD00798296
LHUYXHJTMVKEOM-MRVPVSSYSA-N
tert-butyl 3s-3-amino-4-hexenoate,tert-butyl 3s-3-aminohex-4-enoate
57375595
tert-butyl (3S)-3-aminohex-4-enoate
CC=CC(CC(=O)OC(C)(C)C)N
Quantity 100mg
Packaging Glass Bottle
Assay Percent Range 97%
Formula Weight 185.27
Chemical Name or Material tert-Butyl (3S)-3-amino-4-hexenoate, 97%

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger