Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-amino-1-piperidinecarboxylate, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
87120-72-7
C10H20N2O2
200.28
MFCD01076201
LZRDHSFPLUWYAX-UHFFFAOYSA-N
4-amino-1-boc-piperidine,1-boc-4-aminopiperidine,n-boc-4-aminopiperidine,n-boc-4-amino-piperidine,4-amino-1-n-boc-piperidine,4-amino-n-boc piperidine,1-boc-4-amino piperidine,1-n-boc-4-aminopiperidine,1-t-butoxycarbonyl-4-aminopiperidine,n-1-boc-4-aminopiperidine
1268291
tert-butyl 4-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)N
87120-72-7
C10H20N2O2
200.28
MFCD01076201
LZRDHSFPLUWYAX-UHFFFAOYSA-N
4-amino-1-boc-piperidine,1-boc-4-aminopiperidine,n-boc-4-aminopiperidine,n-boc-4-amino-piperidine,4-amino-1-n-boc-piperidine,4-amino-n-boc piperidine,1-boc-4-amino piperidine,1-n-boc-4-aminopiperidine,1-t-butoxycarbonyl-4-aminopiperidine,n-1-boc-4-aminopiperidine
1268291
tert-butyl 4-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)N
Color Beige to White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Assay Percent Range 97.5% min. (GC)
Formula Weight 200.28
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material tert-Butyl 4-amino-1-piperidinecarboxylate

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger