Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl 4-aminobutyrate hydrochloride, 97%

Click to view available options
Quantity:
1g
5g
58640-01-0
C8H18ClNO2
195.687
MFCD00270592
CZVNLCOJFFYZPG-UHFFFAOYSA-N
tert-butyl 4-aminobutanoate hydrochloride,h-gamma-abu-otbu.hcl,h-gamma-abu-otbu hcl,gamma-aminobutyric acid t-butyl ester hcl,gamma-aminobutyric acid t-butyl ester hydrochloride,4-aminobutyric acid tert-butyl ester hydrochloride,h-,a-abu-otbu.hcl,h-gaba-otbu . hcl,h-.gamma.-abu-otbu.hcl,h-gamma-abu-otbu hydrochloride
24750478
tert-butyl 4-aminobutanoate;hydrochloride
CC(C)(C)OC(=O)CCCN.Cl
58640-01-0
C8H18ClNO2
195.687
MFCD00270592
CZVNLCOJFFYZPG-UHFFFAOYSA-N
tert-butyl 4-aminobutanoate hydrochloride,h-gamma-abu-otbu.hcl,h-gamma-abu-otbu hcl,gamma-aminobutyric acid t-butyl ester hcl,gamma-aminobutyric acid t-butyl ester hydrochloride,4-aminobutyric acid tert-butyl ester hydrochloride,h-,a-abu-otbu.hcl,h-gaba-otbu . hcl,h-.gamma.-abu-otbu.hcl,h-gamma-abu-otbu hydrochloride
24750478
tert-butyl 4-aminobutanoate;hydrochloride
CC(C)(C)OC(=O)CCCN.Cl
Applications
Employed as an important intermediate for raw material for pharmaceutical.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Color White
Quantity 5g
Linear Formula H2NCH2CH2CH2CO2C(CH3)3·HCl
Formula Weight 195.69
Physical Form Powder
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-aminobutyrate hydrochloride

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C