Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate, 95+%, Maybridge™

Click to view available options
Quantity:
10g
Packaging:
Amber Glass Bottle
214834-18-1
C11H20N2O2S
244.353
SCGQNJHAAYUQOO-UHFFFAOYSA-N
1-boc-4-thiocarbamoylpiperidine,tert-butyl 4-aminocarbothioyl tetrahydropyridine-1 2h-carboxylate,1-tert-butoxycarbonylpiperidine-4-carbothioamide,tert-butyl 4-thiocarbamoylpiperidine-1-carboxylate,tert-butyl 4-aminocarbonothioyl piperidine-1-carboxylate,4-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester,piperidine-4-thiocarboxamide, n1-boc protected,1-tert-butoxycarbonyl-4-carbamothioylpiperidine,1-tert-butoxycarbonyl piperidine-4-thiocarboxamide,tert-butyl 4-thiocarbamoyl-piperidine-1-carboxylate
2735648
tert-butyl 4-carbamothioylpiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C(=S)N
214834-18-1
C11H20N2O2S
244.353
SCGQNJHAAYUQOO-UHFFFAOYSA-N
1-boc-4-thiocarbamoylpiperidine,tert-butyl 4-aminocarbothioyl tetrahydropyridine-1 2h-carboxylate,1-tert-butoxycarbonylpiperidine-4-carbothioamide,tert-butyl 4-thiocarbamoylpiperidine-1-carboxylate,tert-butyl 4-aminocarbonothioyl piperidine-1-carboxylate,4-thiocarbamoyl-piperidine-1-carboxylic acid tert-butyl ester,piperidine-4-thiocarboxamide, n1-boc protected,1-tert-butoxycarbonyl-4-carbamothioylpiperidine,1-tert-butoxycarbonyl piperidine-4-thiocarboxamide,tert-butyl 4-thiocarbamoyl-piperidine-1-carboxylate
2735648
tert-butyl 4-carbamothioylpiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C(=S)N
Packaging Amber Glass Bottle
Quantity 10g
Assay Percent Range 95%
Formula Weight 244.36
Percent Purity 95%
Chemical Name or Material tert-Butyl 4-(aminocarbothioyl)tetrahydropyridine-1(2H)-carboxylate