Alfa Aesar™ tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate, 97%

Manufacturer: Alfa Aesar™ H54100MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH54100MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 114772-40-6
    Molecular Formula C18H19BrO2
    Formula Weight 347.24
    MDL Number MFCD06657561
    Synonym 4'-(Bromomethyl)biphenyl-2-carboxylic acid tert-butyl ester
    Chemical Name or Material tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate

    Specifications

    Formula Weight 347.24
    Molecular Formula C18H19BrO2
    MDL Number MFCD06657561
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 250mg
    CAS 114772-40-6
    Synonym 4'-(Bromomethyl)biphenyl-2-carboxylic acid tert-butyl ester
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Chemical Name or Material tert-Butyl 4'-(bromomethyl)biphenyl-2-carboxylate
    Assay 97%

    Description

    Applications
    1,1-Dimethylethyl Esterbiphenyl is used in the preparation of angiotensin II receptor antagonists.

    Solubility
    Soluble in chloroform, ethyl acetate, and DMSO.

    Notes
    Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit