Alfa Aesar™ tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 41857M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA41857M1

 N/A
    N/A

This unit of measure is invalid. 


    Chemical Identifiers

    CAS 75-65-0
    Molecular Formula (CH3)3COH
    Linear Formula (CH3)3COH
    Formula Weight 74.12
    MDL Number MFCD00004464
    UN Number UN1120
    Beilstein 906698
    Merck Index 14,1542
    Synonym 2-Methyl-2-propanol; tert-Butanol
    Chemical Name or Material tert-Butyl alcohol, Anhydrous

    Specifications

    Formula Weight 74.12
    Assay Percent Range 99.8%
    Molecular Formula (CH3)3COH
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula (CH3)3COH
    Merck Index 14,1542
    Health Hazard 1 Danger
    Melting Point 25°C
    UN Number UN1120
    Packaging ChemSeal™ bottles
    Refractive Index 1.3870
    Storage Note 1
    • Packaged under argon
    Color Colorless
    MDL Number MFCD00004464
    Beilstein 906698
    Flash Point 11° (52°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS
    Density 0.786g/mL
    Physical Form Liquid
    CAS 75-65-0
    Synonym 2-Methyl-2-propanol; tert-Butanol
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H225-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 83°C
    Quantity 4 x 1L
    RTECS Number EO1925000
    Chemical Name or Material tert-Butyl alcohol, Anhydrous
    Assay 99.5%

    GHS H Statement
    H225-H332-H319-H335
    Highly flammable liquid and vapor.
    Harmful if inhaled.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit