Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Click to view available options
Quantity:
500mL
4L
Packaging:
HDPE Pressure Bottom Poly Bottle
Poly Bottle
Specifications
Flash Point | 11.00°C |
Quantity | 500mL |
Packaging | HDPE Pressure Bottom Poly Bottle |
Specific Gravity | 1L = 0.78kg |
Formula Weight | 74.12 |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | tert-Butyl Alcohol |