Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl Alcohol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
HDPE Pressure Bottom Poly Bottle
Poly Bottle
75-65-0
C4H10O
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O
75-65-0
C4H10O
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O

High quality chemicals for laboratory and specialized industrial use

Packaging Poly Bottle
Specific Gravity 1L = 0.78kg
Formula Weight 74.12
Flash Point 11.00°C
Quantity 4L
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material tert-Butyl Alcohol