Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl disulfide, 98%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Glass bottle
110-06-5
C8H18S2
178.35
BKCNDTDWDGQHSD-UHFFFAOYSA-N
di-tert-butyl disulfide,disulfide, bis 1,1-dimethylethyl,1,2-di-tert-butyldisulfane,di-tert-butyl disulphide,tert-butyl disulfide,t-butyl disulfide,2-tert-butyldisulfanyl-2-methylpropane,di-t-butyl disulfide,t-butyldisulfide,2,2'-dithiobis 2-methylpropane
66060
2-(tert-butyldisulfanyl)-2-methylpropane
CC(C)(C)SSC(C)(C)C
110-06-5
C8H18S2
178.35
BKCNDTDWDGQHSD-UHFFFAOYSA-N
di-tert-butyl disulfide,disulfide, bis 1,1-dimethylethyl,1,2-di-tert-butyldisulfane,di-tert-butyl disulphide,tert-butyl disulfide,t-butyl disulfide,2-tert-butyldisulfanyl-2-methylpropane,di-t-butyl disulfide,t-butyldisulfide,2,2'-dithiobis 2-methylpropane
66060
2-(tert-butyldisulfanyl)-2-methylpropane
CC(C)(C)SSC(C)(C)C
Specific Gravity 0.93
Color Yellow
Melting Point -5.0°C
Infrared Spectrum Authentic
Flash Point 67°C
Quantity 2.5kg
Refractive Index 1.4882 to 1.4902
Water 0.02% Max. (K.F.)
Packaging Glass bottle
Boiling Point 198.0°C to 204.0°C
Density 0.9300g/mL
Assay Percent Range 97.5% min. (GC)
Linear Formula [(CH3)3C]2S2
Beilstein 01,IV,1638
Solubility Information Solubility in water: insoluble
Formula Weight 178.35
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material tert-Butyl disulfide
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/

GHS Signal Word: Danger

EINECSNumber : 203-734-1