Alfa Aesar™ tert-Butyl ethyl ether, 99%

Manufacturer: Alfa Aesar™ B2294506

Quantity 5g

 View more versions of this product

Catalog No. AAB2294506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 637-92-3
    Molecular Formula (CH3)3COCH2CH3
    Formula Weight 102.18
    MDL Number MFCD00009225
    UN Number UN1179
    Beilstein 1731469
    Merck Index 14,3774
    Synonym Ethyl tert-butyl ether; tert-Butoxyethane
    Chemical Name or Material tert-Butyl ethyl ether

    Specifications

    Formula Weight 102.18
    TSCA TSCA
    Boiling Point 72° to 73°C
    Quantity 5g
    Flash Point −19°C (−2°F)
    Molecular Formula (CH3)3COCH2CH3
    MDL Number MFCD00009225
    CAS 637-92-3
    UN Number UN1179
    Beilstein 1731469
    Merck Index 14,3774
    Synonym Ethyl tert-butyl ether; tert-Butoxyethane
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point −94°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H331-H315-H319-H335
    Highly flammable liquid and vapour.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL BUTYL ETHER
    RTECS Number KN4730200
    Chemical Name or Material tert-Butyl ethyl ether
    Assay 99%

    Description

    Applications
    tert-Butyl ethyl ether is used as an oxygenate gasoline additive oxygenate during its production from crude oil. It is used as an extractant in human urine by using single-walled carbon nanotubes as an adsorbent. It plays an important role as a fuel component in petrol to enhance its octane rating.

    Solubility
    Miscible with alcohol, ethyl ether. Slightly miscible with water.

    Notes
    Incompatible with strong oxidizing agents, neoprene, n-butyl and urethane. It may react with air to form peroxides.

    GHS H Statement
    H225-H331-H315-H319-H335
    Highly flammable liquid and vapour.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit