Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl ethyl malonate, 95%

Manufacturer:  Alfa Aesar™ A1192706

Quantity 5g

 View more versions of this product

Catalog No. AAA1192706


  Check Availability
Add to cart

Specifications

Description

Applications
tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

Solubility
Miscible with most common solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

32864-38-3
188.22
1776681
tert-Butyl ethyl malonate
C9H16O4
MFCD00009193
Malonic acid tert-butyl ethyl ester

Specifications

32864-38-3
188.22
1776681
98° to 100°C (22mmHg)
5g
95%
C9H16O4
MFCD00009193
Malonic acid tert-butyl ethyl ester
87°C (188°F)
tert-Butyl ethyl malonate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit