Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl ethyl malonate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
25g
Packaging:
Glass bottle
32864-38-3
C9H16O4
188.22
MFCD00009193
OCOBFMZGRJOSOU-UHFFFAOYSA-N
tert-butyl ethyl malonate,t-butyl ethyl malonate,propanedioic acid, 1,1-dimethylethyl ethyl ester,1-tert-butyl 3-ethyl propanedioate,tert-butyl ethyl propanedioate,malonic acid tert-butyl ethyl ester,tert-butyl ethyl propane-1,3-dioate,zlchem 692,acmc-1cjju,1-tert-butyl ethyl malonate
96345
3-O-tert-butyl 1-O-ethyl propanedioate
CCOC(=O)CC(=O)OC(C)(C)C
32864-38-3
C9H16O4
188.22
MFCD00009193
OCOBFMZGRJOSOU-UHFFFAOYSA-N
tert-butyl ethyl malonate,t-butyl ethyl malonate,propanedioic acid, 1,1-dimethylethyl ethyl ester,1-tert-butyl 3-ethyl propanedioate,tert-butyl ethyl propanedioate,malonic acid tert-butyl ethyl ester,tert-butyl ethyl propane-1,3-dioate,zlchem 692,acmc-1cjju,1-tert-butyl ethyl malonate
96345
3-O-tert-butyl 1-O-ethyl propanedioate
CCOC(=O)CC(=O)OC(C)(C)C
Specific Gravity 1.001
Flash Point 87°C
Boiling Point 106.0°C to 110.0°C
Density 1.0010g/mL
Refractive Index 1.4163
Packaging Glass bottle
Quantity 25g
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)3CO2CCH2CO2C2H5
Solubility Information Solubility in water: insoluble (20°C). Other solubilities: soluble in most common solvents
Formula Weight 188.22
Percent Purity 97%
Chemical Name or Material tert-Butyl ethyl malonate
Show More Show Less

EINECSNumber : 251-269-8