Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl hydrogen succinate, 97%

Click to view available options
Quantity:
1g
5g
15026-17-2
C8H14O4
174.196
MFCD00273441
PCOCFIOYWNCGBM-UHFFFAOYSA-N
mono-tert-butyl succinate,4-tert-butoxy-4-oxobutanoic acid,succinic acid mono-tert-butyl ester,4-tert-butoxy-4-oxobutanoic acid,tert-butyl hydrogen succinate,mono-t-butyl succinate,pubchem11616,mono-tert-butyl succinate,succinic acid 1-tert-butyl ester,succinic acid 4-tert-butyl ester
4139107
4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
CC(C)(C)OC(=O)CCC(=O)O
15026-17-2
C8H14O4
174.196
MFCD00273441
PCOCFIOYWNCGBM-UHFFFAOYSA-N
mono-tert-butyl succinate,4-tert-butoxy-4-oxobutanoic acid,succinic acid mono-tert-butyl ester,4-tert-butoxy-4-oxobutanoic acid,tert-butyl hydrogen succinate,mono-t-butyl succinate,pubchem11616,mono-tert-butyl succinate,succinic acid 1-tert-butyl ester,succinic acid 4-tert-butyl ester
4139107
4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
CC(C)(C)OC(=O)CCC(=O)O
Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Color Brown-Yellow
Melting Point 51°C to 54°C (literature)
Quantity 5g
Linear Formula HOOCCH2CH2CO2C(CH3)3
Beilstein 1772169
Formula Weight 174.2
Physical Form Solid
Percent Purity 97%
Chemical Name or Material tert-Butyl hydrogen succinate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning