Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl hydroperoxide, 70% Solution in water, ACROS Organics™

Click to view available options
Quantity:
5g
250g
500g
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
75-91-2,7732-18-5
C4H10O2
90.12
CIHOLLKRGTVIJN-UHFFFAOYSA-N
tert-butyl hydroperoxide,tbhp,t-butyl hydroperoxide,tert-butylhydroperoxide,perbutyl h,t-butylhydroperoxide,cadox tbh,hydroperoxide, 1,1-dimethylethyl,1,1-dimethylethyl hydroperoxide,terc. butylhydroperoxid
6410
CHEBI:64090
2-hydroperoxy-2-methylpropane
CC(C)(C)OO
75-91-2,7732-18-5
C4H10O2
90.12
CIHOLLKRGTVIJN-UHFFFAOYSA-N
tert-butyl hydroperoxide,tbhp,t-butyl hydroperoxide,tert-butylhydroperoxide,perbutyl h,t-butylhydroperoxide,cadox tbh,hydroperoxide, 1,1-dimethylethyl,1,1-dimethylethyl hydroperoxide,terc. butylhydroperoxid
6410
CHEBI:64090
2-hydroperoxy-2-methylpropane
CC(C)(C)OO
Specific Gravity 0.94
Viscosity 4.1 mPa.s (20°C)
Refractive Index 1.3860 to 1.3880
Quantity 2.5kg
Color Colorless
Density 0.9400g/mL
Melting Point -2.8°C
Boiling Point 37.0°C (15.0 mmHg)
Packaging Plastic bottle
Flash Point 43°C
Linear Formula (CH3)3COOH
Beilstein 01,IV,1616
Fieser 01,88; 02,49; 03,37; 05,75; 06,81; 07,43; 08,62; 09,78; 10,64; 11,91; 12,88; 14,60; 15,55; 16,53; 17,56
Merck Index 15,1573
Solubility Information Solubility in water: miscible. Other solubilities: soluble in hydroxide solutions,soluble in most common organic solvents
Formula Weight 90.12
Physical Form Liquid
Chemical Name or Material tert-Butyl hydroperoxide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 200-915-7