Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, 99.9%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
Acetic Acid 0.005% max.
Additional Information TBA content < 0.05%
Assay Percent Range 99.9%
Linear Formula (CH3)3COCH3
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Formula Weight 88.15
Physical Form Liquid
Percent Purity 99.9%
Grade Extra Pure
Density 0.7404g/mL
Methanol 0.05% max.
Packaging Glass bottle
Peroxides 1mg/kg H2O2 max.
Refractive Index 1.3675 to 1.3695
Residue after Evaporation 1mg/100mL max.
Solubility Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Specific Gravity 0.7404
Water 0.01% max.
Flash Point −28°C
Viscosity 0.36 mPa.s (20°C)
Boiling Point 54.0°C to 56.0°C
Color Undesignated
Melting Point -110.0°C
Quantity 1L
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less

GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger