Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, 99%, pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
25L
Packaging:
Glass bottle
Metal Drum
Plastic drum
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
Assay Percent Range 99%
Linear Formula (CH3)3COCH3
Beilstein 01, 381
Fieser 04,333
Merck Index 15, 6106
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Formula Weight 88.15
Physical Form Powder or Needles
Percent Purity 99%
Grade Pure
Density 0.7404g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3675 to 1.3695
Solubility Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Specific Gravity 0.7404
Water 0.01% Max. (Coulometric)
Flash Point −28°C
Viscosity 0.36 mPa.s (20°C)
Boiling Point 54.0°C to 56.0°C
Color White to Yellow
Melting Point -110.0°C
Quantity 2.5L
Chemical Name or Material tert-Butyl methyl ether, 99%
Show More Show Less

GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger