Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl methyl ether, HPLC Grade, 99+%

Chromatography solvent

Manufacturer:  Alfa Aesar™ 40477M6

Quantity 4 x 4L

 View more versions of this product

Catalog No. AA40477M6


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1634-04-4
MFCD00008812
1730942
Methyl tert-butyl ether
HPLC Grade
88.15
UN2350
14,6032
tert-Butyl methyl ether

Specifications

1634-04-4
88.15
UN2350
14,6032
0.742g/mL
1.3690
55° to 56°C
4 x 4L
≥99%
Liquid
MFCD00008812
1730942
Methyl tert-butyl ether
Mild
−25° (−13°F)
−109°C
tert-Butyl methyl ether
HPLC Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H315
Highly flammable liquid and vapor.
Causes skin irritation.

GHS P Statement
P210-P280-P303+P361+P353-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL METHYL ETHER

RTECSNumber : KN5250000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit