Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert.-Butyl methyl malonate, 95%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
25mL
Packaging:
Glass bottle
42726-73-8
C8H14O4
174.2
MFCD00042856
XPSYZCWYRWHVCC-UHFFFAOYSA-N
tert-butyl methyl malonate,t-butyl methyl malonate,1-tert-butyl 3-methyl propanedioate,acmc-20ajkv,tert-butyl methylmalote,pubchem20476,methyl tert-butyl malonate,ksc493c8h,tert-butyl methyl malonaoate
2733872
3-O-tert-butyl 1-O-methyl propanedioate
CC(C)(C)OC(=O)CC(=O)OC
42726-73-8
C8H14O4
174.2
MFCD00042856
XPSYZCWYRWHVCC-UHFFFAOYSA-N
tert-butyl methyl malonate,t-butyl methyl malonate,1-tert-butyl 3-methyl propanedioate,acmc-20ajkv,tert-butyl methylmalote,pubchem20476,methyl tert-butyl malonate,ksc493c8h,tert-butyl methyl malonaoate
2733872
3-O-tert-butyl 1-O-methyl propanedioate
CC(C)(C)OC(=O)CC(=O)OC
Flash Point 79°C
Melting Point -10°C
Boiling Point 80°C (11.0mmHg)
Packaging Glass bottle
Refractive Index 1.4140 to 1.417
Specific Gravity 1.03
Density 1.0300g/mL
Infrared Spectrum Authentic
Quantity 1mL
Color Colorless to Yellow
Assay Percent Range 94% min. (GC)
Linear Formula H3COOCCH2COOC(CH3)3
Formula Weight 174.2
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material tert.-Butyl methyl malonate
Show More Show Less

EINECSNumber : 255-919-1

Recommended Storage : Product may discolor to yellow on storage