Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl methyl sulfide, 98+%

Click to view available options
Quantity:
5g
25g
6163-64-0
C5H12S
104.211
MFCD00008840
CJFVCTVYZFTORU-UHFFFAOYSA-N
tert-butyl methyl sulfide,propane, 2-methyl-2-methylthio,sulfide, tert-butyl methyl,methyl tert-butyl sulfide,3,3-dimethyl-2-thiabutane,2-methyl-2-methylsulfanyl propane,t-butyl methyl sulfide,methyl t-butyl sulfide,tert-butyl methyl sulfane,acmc-1b7tx
138679
2-methyl-2-methylsulfanylpropane
CC(C)(C)SC
6163-64-0
C5H12S
104.211
MFCD00008840
CJFVCTVYZFTORU-UHFFFAOYSA-N
tert-butyl methyl sulfide,propane, 2-methyl-2-methylthio,sulfide, tert-butyl methyl,methyl tert-butyl sulfide,3,3-dimethyl-2-thiabutane,2-methyl-2-methylsulfanyl propane,t-butyl methyl sulfide,methyl t-butyl sulfide,tert-butyl methyl sulfane,acmc-1b7tx
138679
2-methyl-2-methylsulfanylpropane
CC(C)(C)SC
Applications
tert-Butyl methyl sulfide is used for LPG refining process and It have high content of sulfur.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 5g
Odor Unpleasant
Flash Point −3°C (26°F)
Boiling Point 101°C to 102°C
Linear Formula (CH3)3CSCH3
UN Number UN1993
Formula Weight 104.22
Percent Purity ≥98%
Chemical Name or Material tert-Butyl methyl sulfide

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P261-P243-P403
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.