Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl methyl ether, for analysis, Acros Organics

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
1634-04-4
C5H12O
88.15
MFCD00008812
BZLVMXJERCGZMT-UHFFFAOYSA-N
tert-butyl methyl ether,methyl tert-butyl ether,mtbe,methyl t-butyl ether,t-butyl methyl ether,methyl tertiary-butyl ether,propane, 2-methoxy-2-methyl,methyl-tert-butyl ether,methyl-t-butyl ether,2-methyl-2-methoxypropane
15413
CHEBI:27642
2-methoxy-2-methylpropane
CC(C)(C)OC
Specific Gravity 0.7404
Water 0.05% max.
Viscosity 0.36 mPa.s (20°C)
Residue after Evaporation 0.005% max.
Packaging Glass bottle
Refractive Index 1.3675 to 1.3695
Quantity 1L
Flash Point −28°C
Infrared Spectrum Authentic
Color Colorless
Melting Point -110.0°C
Density 0.7404g/mL
Acidity 0.0005 meq/g max.
Boiling Point 54.0°C to 56.0°C
Linear Formula (CH3)3COCH3
Merck Index 15, 6106
Solubility Information Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
Formula Weight 88.15
Physical Form Liquid
Percent Purity ≥99.5%
Grade Analysis
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less

GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.

GHS Signal Word: Danger

EINECSNumber : 216-653-1