Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl N-hydroxycarbamate, 98+%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
36016-38-3
C5H11NO3
133.15
MFCD00002107
DRDVJQOGFWAVLH-UHFFFAOYSA-N
n-boc-hydroxylamine,n-tert-butoxycarbonylhydroxylamine,tert-butyl hydroxycarbamate,boc-amino alcohol,carbamic acid, hydroxy-, 1,1-dimethylethyl ester,n-hydroxycarbamic acid tert-butyl ester,tert-butyl-n-hydroxycarbamate,n-tert-boc-hydroxylamine,t-butyl n-hydroxycarbamate,n-tert-butyloxycarbonylhydroxylamine
97534
tert-butyl N-hydroxycarbamate
CC(C)(C)OC(=O)NO
36016-38-3
C5H11NO3
133.15
MFCD00002107
DRDVJQOGFWAVLH-UHFFFAOYSA-N
n-boc-hydroxylamine,n-tert-butoxycarbonylhydroxylamine,tert-butyl hydroxycarbamate,boc-amino alcohol,carbamic acid, hydroxy-, 1,1-dimethylethyl ester,n-hydroxycarbamic acid tert-butyl ester,tert-butyl-n-hydroxycarbamate,n-tert-boc-hydroxylamine,t-butyl n-hydroxycarbamate,n-tert-butyloxycarbonylhydroxylamine
97534
tert-butyl N-hydroxycarbamate
CC(C)(C)OC(=O)NO
Packaging Glass bottle
Infrared Spectrum Authentic
Color Pink to White
Quantity 1g
Melting Point 53.0°C to 57.0°C
Assay Percent Range 98% min. (GC)
Linear Formula HONHCO2C(CH3)3
Beilstein 03,IV,168
Formula Weight 133.15
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material tert-Butyl N-hydroxycarbamate, 98+%
Show More Show Less

MOISTURE SENSITIVE

EINECSNumber : 252-836-2