Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl N-hydroxycarbamate, 98+%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Color | Pink to White |
Melting Point | 53.0°C to 57.0°C |
Quantity | 25g |
Assay Percent Range | 98% min. (GC) |
Linear Formula | HONHCO2C(CH3)3 |
Beilstein | 03,IV,168 |
Formula Weight | 133.15 |
Physical Form | Crystalline Powder |
Show More |
Safety and Handling
MOISTURE SENSITIVE
EINECSNumber : 252-836-2