Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl nitrite, 90%, pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic drum
540-80-7
C4H9NO2
103.12
IOGXOCVLYRDXLW-UHFFFAOYSA-N
t-butyl nitrite,1,1-dimethylethyl nitrite,tert-butylnitrite,nitrous acid, 1,1-dimethylethyl ester,unii-4780h7u8lu,ccris 2098,alpha,alpha-dimethylethyl nitrite,nitrous acid, t-butyl ester,2-methyl-2-propylnitrit,ch3 3cono
10906
tert-butyl nitrite
CC(C)(C)ON=O
540-80-7
C4H9NO2
103.12
IOGXOCVLYRDXLW-UHFFFAOYSA-N
t-butyl nitrite,1,1-dimethylethyl nitrite,tert-butylnitrite,nitrous acid, 1,1-dimethylethyl ester,unii-4780h7u8lu,ccris 2098,alpha,alpha-dimethylethyl nitrite,nitrous acid, t-butyl ester,2-methyl-2-propylnitrit,ch3 3cono
10906
tert-butyl nitrite
CC(C)(C)ON=O
Specific Gravity 0.86
Packaging Plastic drum
Refractive Index 1.3670 to 1.3700
Flash Point −10°C
Density 0.8600g/mL
Color Yellow
Boiling Point 61.0°C to 63.0°C
Quantity 2.5kg
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Linear Formula (CH3)3CNO2
Beilstein 01,382
Fieser 07,48
Merck Index 15,1586
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,chloroform and carbon,disulfide,practically insoluble in glycerol
Formula Weight 103.12
Physical Form Liquid
Percent Purity 90%
Grade Pure
Chemical Name or Material tert-Butyl nitrite
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 208-757-