Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl peroxyacetate, 50% solution in aromatic free mineral spirit, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiators
Specifications
Color | Undesignated |
Formula Weight | 132.16 |
Percent Purity | 49 to 51% |
Packaging | Plastic bottle |
Quantity | 250 mL |
Physical Form | Solution |
Chemical Name or Material | tert-Butyl peroxyacetate, 50% Solution In Aromatic Free Mineral Spirit |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement Flammable liquid and vapour. Heating may cause a fire. Causes serious eye irritation. May cause an allergic skin reaction. May be fatal if swallowed and enters airways. Toxic if inhaled. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction