Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl phenyl carbonate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
6627-89-0
C11H14O3
194.23
UXWVQHXKKOGTSY-UHFFFAOYSA-N
t-butyl phenyl carbonate,tert-butylphenylcarbonate,2-methyl-2-propanyl phenyl carbonate,carbonic acid, 1,1-dimethylethyl phenyl ester,pubchem13300,acmc-209nuo,phenyl t-butyl carbonate,uxwvqhxkkogtsy-uhfffaoysa,tert-butyl phenyl carbonate,carbonic acid tert-butylphenyl ester
81113
tert-butyl phenyl carbonate
CC(C)(C)OC(=O)OC1=CC=CC=C1
6627-89-0
C11H14O3
194.23
UXWVQHXKKOGTSY-UHFFFAOYSA-N
t-butyl phenyl carbonate,tert-butylphenylcarbonate,2-methyl-2-propanyl phenyl carbonate,carbonic acid, 1,1-dimethylethyl phenyl ester,pubchem13300,acmc-209nuo,phenyl t-butyl carbonate,uxwvqhxkkogtsy-uhfffaoysa,tert-butyl phenyl carbonate,carbonic acid tert-butylphenyl ester
81113
tert-butyl phenyl carbonate
CC(C)(C)OC(=O)OC1=CC=CC=C1
Refractive Index 1.4805
Boiling Point 83.0°C (0.6 mmHg)
Specific Gravity 1.05
Quantity 100g
Color Colorless
Flash Point 101°C
Density 1.0500g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5OCO2C(CH3)3
Fieser 05,86
Formula Weight 194.23
Physical Form Liquid or Low Melting Solid
Percent Purity 98%
Chemical Name or Material tert-Butyl phenyl carbonate
Show More Show Less

EINECSNumber : 229-601-8