Alfa Aesar™ tert-Butyl phenyl ether, 98%

Manufacturer: Alfa Aesar™ H27689MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27689MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6669-13-2
    Molecular Formula C10H14O
    Formula Weight 150.22
    MDL Number MFCD00049018
    Synonym tert-Butoxybenzene
    Chemical Name or Material tert-Butyl phenyl ether

    Specifications

    Formula Weight 150.22
    Molecular Formula C10H14O
    MDL Number MFCD00049018
    CAS 6669-13-2
    Synonym tert-Butoxybenzene
    Boiling Point 184°C
    Quantity 250mg
    Chemical Name or Material tert-Butyl phenyl ether
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit