Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ tert-Butyl phenyl ether, 98%

Manufacturer:  Alfa Aesar™ H2768906

Quantity 5g

 View more versions of this product

Catalog No. AAH2768906


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6669-13-2
150.22
tert-Butoxybenzene
C10H14O
MFCD00049018
tert-Butyl phenyl ether

Specifications

6669-13-2
150.22
tert-Butoxybenzene
5g
98%
C10H14O
MFCD00049018
184°C
tert-Butyl phenyl ether
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit