Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
tert-Butyl pivalate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
tert-Butyl pivalate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl pivalate is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
tert-Butyl pivalate is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Density | 0.813 |
Boiling Point | 134°C to 135°C |
Flash Point | >62°C (144°F) |
Refractive Index | 1.392 |
Linear Formula | (CH3)3CCO2C(CH3)3 |
Quantity | 5 g |
Synonym | tert-butyl pivalate, tert-butyl trimethylacetate, propanoic acid, 2,2-dimethyl, 1,1-dimethylethyl ester, di-tert-butylcarboxylic acid, 2,2-dimethylpropanoic acid tert-butyl ester, 2,2-dimethyl-propionic acid tert-butyl ester, propanoic acid, 2,2-dimethyl-, 1,1-dimethylethyl ester, propanoic acid,2,2-dimethyl-, 1,1-dimethylethyl ester |
Formula Weight | 158.24 |
Percent Purity | 98% |
Chemical Name or Material | tert-Butyl pivalate |
Safety and Handling
Hazard Category | H227 |
Hazard Statement | GHS H Statement H227 Combustible liquid. |
Precautionary Statement | P210-P280-P370+P378q-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction